Mantle for Squibb-Type Funnels, Ace Glass Incorporated
Supplier: Ace Glass
12038-04 12038-06 12038-08 12038-10
89075-912EA 1028.11 USD
89075-912 89075-914 89075-916 89075-918
Mantle for Squibb-Type Funnels, Ace Glass Incorporated
Heaters Heating Mantles
Designed for use with Squibb type separatory funnels
Mantles are supplied with separable 122 cm (4'), 2-wire cord, and locking connector. Large sizes of one liter and up have viewing slots, as shown.
Certificaciones: CSA approved.