Flexible Motor Drive Shaft, Ace Glass Incorporated
Supplier: Ace Glass
8081-05 8081-06 8081-07 8081-08 8081-12 8081-15 8081-19 8081-24 8081-27 8081-30 8081-32
89062-032EA 100 USD
89062-032 89062-034 89062-036 89062-038 89062-040 89062-042 89062-044 89062-046 89062-048 89062-050 89062-052
Flexible Motor Drive Shaft, Ace Glass Incorporated
Stirrers Stirring Shafts
Fully flexible drive shaft connects your stirring motor to any size or type of reactor stir shaft
The shafts operate up to 14,000 rpm, and they should be run in a counter-clockwise direction. Typical torque ratings — sharp bend in shaft, (4-inch loop): 4.7 Kg-cm, (4 in-lbs); Straight shaft: 28 Kg-cm, (24 in-lbs). Shafts measure approximately 91.4 cm, (36 inches) or 152.4 cm, (60 inches) with hand-piece and motor coupling attached.
Designed with a ball bearing motor coupling at one end for connection to any motor with an 8mm, (5/16-inch) diameter shaft. The other end has a detachableMore Product Information