Air Sampling Cane with Funnel, Ace Glass Incorporated
Supplier: Ace Glass
7493-12 7493-16 7493-22 7493-26
89059-174EA 876.28 USD
89059-174 89059-176 89083-166 89083-168
Air Sampling Cane with Funnel, Ace Glass Incorporated
Manifolds
The air sampling cane is fabricated from borosilicate glass.
The air sampling cane is equipped with a funnel at one end to prevent water droplets from collecting at the edge and being sucked into the manifold train. A bulge ("W") located above the funnel is designed for inserting glass wool (or similar material) to allow for filtering of particulates. This piece is normally installed outside the van, and is connected at the roof with a roof attachment. Inside the van, it is then connected to the manifold train with a bush with the Size 25 or 50.
Información para pedidos: Available withMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable