Reaction Bottles for Hydrogenation/Gas Apparatus, Ace Glass Incorporated
Supplier: Ace Glass
7478-05 7478-10 7478-12 7478-15 7478-34 7478-37
89058-836EA 90.52 USD
89058-836 89058-838 89058-840 89058-842 89058-844 89058-846
Reaction Bottles for Hydrogenation/Gas Apparatus, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
These borosilicate glass bottles have a tooled neck to accept a #6 rubber stopper
They are epoxy-coated to help prevent scratching. Bottles are rated at 60psi @ 20°C. Working capacity is ½ the actual capacity listed below.
Información para pedidos: Bottles and stoppers sold separately.