Sample Tube, Ace Glass Incorporated
Supplier: Ace Glass
7410-10
89058-818EA 359.19 USD
89058-818
Sample Tube, Ace Glass Incorporated
Tubes Test Tubes
Designed by Dr. H.R. Krouse, University of Calgary, for use with H₂O¹⁸/H₂O¹⁶ determinations.
CO₂ from the equilibration flask is transferred to the sample tube for isotopic analysis. Unit consists of PTFE plug with FETFE O-Ring and glass vessel, with 5 ml tube at bottom.