Flask, Rotary Evaporator/Freeze Drying, Ace Glass Incorporated
Supplier: Ace Glass
7030-03 7030-04 7030-06 7030-08 7030-10 7030-12 7030-14 7030-21 7030-23 7030-27
89057-464EA 334.5 USD
89057-464 89057-466 89057-468 89057-470 89057-472 89057-474 89057-476 89057-478 89057-480 89057-482
Flask, Rotary Evaporator/Freeze Drying, Ace Glass Incorporated
Flasks Rotary Evaporator Flasks
This a dual purpose flask features the “TWISTLOR” ACE-Seal: simply push the adapter head and the flask together and turn 90°
To remove, turn 90° in either direction, or until the nylon retainers line up with flats on the flange and the flask and adapter head come apart. Fabricated from borosilicate glass.
The flange on flask has two ground-flats opposite each other that allow nylon retainers on the adapter head to pass over. An o-ring positioned in grooves on the head and flask makes a vacuum- tight seal when the head is turned 90° in either direction.
Información para pedidos: More Product Information