Kuderna-Danish Evaporative Concentrator, with Springs, Ace Glass Incorporated
Supplier: Ace Glass
6708-03 6708-11 6708-13 6708-15 6708-17 6708-35 6708-37 6708-38
89055-894EA 165.69 USD
89055-894 89055-902 89055-904 89055-906 89055-908 89055-918 89055-920 89055-922
Kuderna-Danish Evaporative Concentrator, with Springs, Ace Glass Incorporated
Concentrators
This Kuderna-Danish evaporative apparatus is used to concentrate materials in volatile solvents
It combines rapidity, simplicity, and quantitative efficiency, including the minimization of entrainment escape and transfer losses. The flask has a [ST] 19/22 inner joint on the bottom, and a reinforced [ST] 24/40 outer joint on top. The flask also comes complete with 89061-668 stainless steel springs used to connect it to the receiver. Receivers have [ST] 19/22 outer joint at top for connection to flask bottom. Column has a [ST] 24/40 reinforced outer joint at top, and a [ST] inner joint atMore Product Information