Reaction Flask with Indents, 4-Inch Conical Flange, Ace Glass Incorporated
Supplier: Ace Glass
6477-05 6477-15 6477-20 6477-10 6477-25
89054-890EA 348.52 USD
89054-890 89054-894 89054-896 89054-902 89054-930
Reaction Flask with Indents, 4-Inch Conical Flange, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Flask Shape
This rugged, cylindrical reaction flask has a conical neck opening of four inches (100 mm)
It has side indents for improved stirring characteristics. Not suitable for vacuum operation. Use a 89055-014 clamp to secure the head to the flask. For gaskets, see 89055-010 (or similar). Fabricated from borosilicate glass.
Información para pedidos: Clamp and gasket not included. Use a 89055-014 clamp to secure the head to the flask. For gaskets, see 89055-004.
Order Now
SPECIFICATIONS
Environmentally Preferable