Desiccator, Plastic, Ace Glass Incorporated
Supplier: Ace Glass
6248-12 6248-15 6248-21 6248-31 6248-35 6248-39
89054-050EA 157.85 USD
89054-050 89054-052 89054-054 89054-056 89054-058 89054-060
Desiccator, Plastic, Ace Glass Incorporated
Desiccators Desiccators, Plastic
This plastic desiccator has a transparent polycarbonate top, and a white polypropylene bottom
Space-saving design gives an average of 13% greater interior volume. Extra heavy walls aid in avoiding danger of implosion. This shatterproof desiccator will hold a vacuum of 740mm of Hg (29") for a period of 24 hours. Three-way polypropylene stopcock barrel with PTFE plug has a vacuum draw that accepts 1/4-inch I.D. tubing. Third port allows turbulence-free vacuum release. Neoprene o-ring seals lid to bottom and does not need grease.
Información para pedidos: Supplied with perforatedMore Product Information