AlphaTec® 02-100 Chemical resistant gloves, Ansell
Supplier: Ansell Healthcare
988828 988825 988827 988826 988829 902209
89041-864CS 1445.07 USD
89041-864 89041-858 89041-860 89041-862 89041-866 89041-856
AlphaTec® 02-100 Chemical resistant gloves, Ansell
Gloves
Style 02-100 flat-film gloves are constructed of a five-layer composite of polyethylene, nylon, and metallocene.
  • Can be worn beneath heavier gloves
  • Useful for handling in many different industries
  • Greater dexterity with winged thumbs
  • Hand-specific
The unique polymer and seam-fusion technology provide a secure seal against a wide range of chemicals, as well as a flexible, comfortable fit.
Uses include aircraft assembly; chemical processing, oil refining and petrochemicals; handling acids, alkalies, salts, ketones and alcohols; handling strongMore Product Information