ART® and Hydrologix (HLT) SoftFit-L™ Pipette Tips for Rainin® LTS Pipettors, Molecular BioProducts
Supplier: Thermo Scientific
2749-HR 2749-05-HR 3721-HR 3721-05-HR 3722-HR 3722-05-HR 3721-RI 3721-05-RI 3722-RI 3722-05-RI 3704-ER 2769-HR 2769-05-HR 3751-HR 3751-05-HR 3752-HR 3752-05-HR 3751-RI 3751-05-RI 3752-RI 3752-05-RI 2739-HR 2739-05-HR 3731-HR 3731-05-HR 3732-HR 3732-05-HR 3731-RI 3731-05-RI 3732-RI 3732-05-RI 2779-HR 2779-05-HR 3781-HR 3781-05-HR 3782-HR 3782-05-HR 3781-RI 3781-05-RI 3782-RI 3782-05-RI 3706-ER 2789-HR 2789-05-HR 3741-HR 3742-HR 3741-RI 3742-RI 3742-05-RI
89031-350CS 1597.35 USD
89031-350 89031-352 89031-354 89031-356 89031-358 89031-360 89031-362 89031-364 89031-366 89031-368 89031-370 89031-372 89031-374 89031-376 89031-378 89031-380 89031-382 89031-384 89031-386 89031-388 89031-390 89031-392 89031-394 89031-396 89031-398 89031-400 89031-402 89031-404 89031-406 89031-408 89031-410 89031-412 89031-414 89031-416 89031-418 89031-420 89031-422 89031-424 89031-426 89031-428 89031-430 89031-432 89031-434 89031-436 89031-438 89031-442 89031-446 89031-450 89031-452
ART® and Hydrologix (HLT) SoftFit-L™ Pipette Tips for Rainin® LTS Pipettors, Molecular BioProducts
Pipette Tips
SoftFit-L™ pipette tips are available in five volume sizes, and are designed for use with Rainin® LTS pipettors.
  • Certified pure
  • Reload inserts are color-coded by size
Tips deliver highly accurate samples, seal well on the pipettor, and are easy to seat and eject. To accommodate a variety of applications, SoftFit-L™ pipette tips offer an expanded selection of ART®, HLT, and low retention designs. The ART® (Aerosol Resistant Tips) series features a self-sealing barrier designed to prevent contamination even under the most demanding conditions.