VWR® Critical Swab® Swab, Cone-Shaped Reticulated Foam Head
Supplier: VWR International
89031-282
89031-282CS 410.24 USD
89031-282
VWR® Critical Swab® Swab, Cone-Shaped Reticulated Foam Head
Swabs Swabs for Sampling
Swab features reticulated foam and a blue glass-filled polypropylene shaft for extra rigidity.
  • Reticulated foam
  • Polypropylene shaft
  • Extra rigid
For use in critical environments.