VWR®, Beakers, PTFE
Supplier: VWR International
89026-010 89026-012 89026-014 89026-016 89026-020 89026-022 89026-024 89026-026 89026-028 89026-030 89026-032 89026-034 89026-036
89026-010EA 462.97 USD
89026-010 89026-012 89026-014 89026-016 89026-020 89026-022 89026-024 89026-026 89026-028 89026-030 89026-032 89026-034 89026-036
VWR®, Beakers, PTFE
Beakers
Isostatically molded in pure, FDA approved PTFE, these beakers feature a pouring spout and have a machined base for good heat transfer.
  • Chemically resistant and inert
  • Autoclavable
Machined base provides good heat transfer in these PTFE beakers.