Surgi-OR™ Graves Vaginal Speculum, Mid Grade, Sklar
Supplier: Sklar
95-303 95-304 95-312 95-322 95-324
89023-558EA 55.7 USD
89023-558 89023-560 89023-564 89023-568 89023-570
Surgi-OR™ Graves Vaginal Speculum, Mid Grade, Sklar
Specula
Surgi-OR™ Graves Vaginal Speculum is a gynecological instrument used for performing pelvic examinations, pap smears, and examining the cervix.
  • Latex-Free
  • It is a double blade duckbill speculum inserted with the blades closed
The blades are then opened by an external lever and screw. The Graves Speculum has wider blades than the Pederson Speculum and the practitioner can best determine which type and size speculum is most appropriate for the patient.
Hand-crafted in Pakistan, Surgi-OR™ mid-grade instruments are forged from high quality JapaneseMore Product Information