VWR® Freestanding Specimen and Centrifuge Tubes, PP, with Plug Seal Caps
Supplier: VWR International
CFT001030 CFT000030 CFT002030
89012-778CS 206.8 USD
89012-778 89012-780 89012-782
VWR® Freestanding Specimen and Centrifuge Tubes, PP, with Plug Seal Caps
Tubes Centrifuge Tubes
Made of polypropylene, these tubes feature molded gradutions and a leak-proof plug seal cap designed to withstand 95 kPa of pressure.
  • Polypropylene fabrication
  • Skirted design
  • Plug seal caps
  • Available sterile or non-sterile
Skirted design enables tubes to stand upright, allowing for use without separate racks or stands.
Envase: Tubes are available in bags of 50 with attached cap, or bulk packaged with 500 separate caps.