Maintenance-Free Tyvek® Hood Assembly with Low Pressure Flow Adapter and Suspension, Allegro
Supplier: Allegro
9910
89011-732EA 151.32 USD
89011-732
Maintenance-Free Tyvek® Hood Assembly with Low Pressure Flow Adapter and Suspension, Allegro
Disposable Garments
Our NIOSH-approved Fully Disposable Tyvek Hood is so unique, it is patented.
  • Poly-Coated DuPont Tyvek Hood
  • Splash-Resistant Hood and Bib
  • Seams Surged to Seal in The Air Flow
  • Generous Size is Ideal for Beards or Goggles
  • Low-Profile, Lightweight Flow Adapter
It’s designed and constructed in one piece with the down tube and head suspension completely integrated
The Low Pressure Flow Adapter allows the hood to connect to an ambient air pump or any low pressure air source. It's economical enough to dispose of with the hood assembly or save and use withMore Product Information