LABSET® Organic Chemistry Glassware Kit, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
29057F-125
89002-134CS 407.77 USD
89002-134 89002-162 26387-165
LABSET® Organic Chemistry Glassware Kit, Kimble Chase
Microchemistry Kits
Glassware kit contains components made of KIMAX® brand borosilicate glass with full scale, full length [ST] 24/40 joints
  • Heavy duty construction and consistent quality
  • Long, trouble-free life
  • Minimized frequency of chipping or breakage in assembly and use
"The kit contains one each of the listed components. The ground joints have a cylindrical outside wall that provides a firm clamping surface for reduced breakage and greater safety. The separatory funnel has reinforced construction, with extra glass provided around the stopcock. The separatory funnelMore Product Information