Kimax® Borosilicate Pertri Dish Covers, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
23062-10010
89001-250CS 91.45 USD
89001-250
Kimax® Borosilicate Pertri Dish Covers, DWK Life Sciences
Culture Dishes Petri Dishes
Replacement glass covers can be used with dishes for culturing bacteria and other general laboratory applications.
  • Covers remain clear after repeated use in wet or dry sterilization cycles
  • Reinforced beaded edges resist mechanical breakage
  • Combination of different colors and wording permits quick and easy identification
  • Manufactured from 33 expansion, low extractable borosilicate glass
Covers have a fused-on bright red ceramic enamel marking and inscription.
Certificaciones: Designed to meet Federal Specification NNN-D-1478. Material conforming toMore Product Information