Acetic acid ≥99%, ARISTAR® ULTRA, Ultrapure for trace metal analysis, VWR Chemicals BDH®

Supplier: BDH
Danger

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

20601-VWNF03
87003-212EA 1205.45 USD
87003-212
Acetic acid ≥99%, ARISTAR® ULTRA, Ultrapure for trace metal analysis, VWR Chemicals BDH®
Acetic acid

Ultra high purity acid for quantitative trace metal analysis at the parts-per-trillion (ppt) level.

Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118
Melting Pt: 17
Flash Pt: 38.5
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay ≥ 99%
Aluminum (Al) max. 50 ppt
Antimony (Sb) max. 50 ppt
Arsenic (As) max. 50 ppt
Barium (Ba) max. 10 ppt
Beryllium (Be) max. 10 ppt
Bismuth (Bi) max. 10 ppt
Cadmium (Cd) max. 10 ppt
Calcium (Ca) max. 50 ppt
Cerium (Ce) max. 10 ppt
Cesium (Cs) max. 10 ppt
Chromium (Cr) max. 10 ppt
Cobalt (Co) max. 10 ppt
Copper (Cu) max. 10 ppt
Dysprosium (Dy) max. 1 ppt
Erbium (Er) max. 1 ppt
Europium (Eu) max. 1 ppt
Gadolinium (Gd) max. 1 ppt
Gallium (Ga) max. 10 ppt
Germanium (Ge) max. 10 ppt
Hafnium (Hf) max. 10 ppt
Holmium (Ho) max. 1 ppt
Indium (In) max. 1 ppt
Iron (Fe) max. 50 ppt
Lanthanum (La) max. 1 ppt
Lead (Pb) max. 10 ppt
Lithium (Li) max. 10 ppt
Lutetium (Lu) max. 10 ppt
Magnesium (Mg) max. 50 ppt
Manganese (Mn) max. 10 ppt
Molybdenum (Mo) max. 10 ppt
Neodymium (Nd) max. 1 ppt
Nickel (Ni) max. 50 ppt
Platinum (Pt) max. 50 ppt
Potassium (K) max. 50 ppt
Praseodymium (Pr) max. 1 ppt
Rhenium (Re) max. 10 ppt
Rhodium (Rh) max. 50 ppt
Rubidium (Rb) max. 10 ppt
Ruthenium (Ru) max. 50 ppt
Samarium (Sm) max. 1 ppt
Scandium (Sc) max. 10 ppt
Selenium (Se) Information only
Silver (Ag) max. 50 ppt
Sodium (Na) max. 100 ppt
Strontium (Sr) max. 10 ppt
Tellurium (Te) max. 1 ppt
Terbium (Tb) max. 1 ppt
Thallium (Tl) max. 10 ppt
Thorium (Th) max. 1 ppt
Thulium (Tm) max. 1 ppt
Tin (Sn) max. 50 ppt
Titanium (Ti) max. 10 ppt
Tungsten (W) max. 10 ppt
Uranium (U) max. 1 ppt
Vanadium (V) max. 10 ppt
Ytterbium (Yb) max. 1 ppt
Yttrium (Y) max. 1 ppt
Zinc (Zn) max. 1 ppt
Zirconium (Zr) max. 10 ppt


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR