UNISETTE™ Processing and Embedding Cassettes, Simport
Supplier: Simport Scientific
M506-11 M505-7 M505-8 M506-2 M506-3 M506-5 M506-7 M506-9 M506-17 M505-4 M505-5 M505-11 M505-12 M505-18 M506-8 M505-2 M505-3 M505-6 M505-9 M505-10 M505-17 M506-4 M506-6 M506-10 M506-12 M506-16 M506-18
87002-336CS 347 USD
87002-336 87002-300 87002-302 87002-318 87002-320 87002-324 87002-328 87002-332 87002-342 87002-294 87002-296 87002-308 87002-310 87002-316 87002-330 87002-290 87002-292 87002-298 87002-304 87002-306 87002-312 87002-314 87002-322 87002-326 87002-334 87002-338 87002-340 87002-344
UNISETTE™ Processing and Embedding Cassettes, Simport
Embedding Cassettes Tissue Processing Cassettes
These cassettes are made of a high-density acetal polymer and keep specimens safely submerged in liquid
The disposable biopsy cassettes have 1mm square openings and are specially designed to hold biopsy specimens during the embedding process, as well as in a storage file.
The cassettes are totally resistant to the chemical action of histological solvents. The flow-through slots allow for maximum fluid exchange and ensure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. The snap-latch and hinge-lock design prevents early separation of base and lidMore Product Information