Polybead® Microspheres, Polysciences
Supplier: Polysciences Inc
07312-5
87001-892EA 694.38 USD
87001-892
Polybead® Microspheres, Polysciences
Beads Microcarriers
Polybead® microspheres are ideally suited for protein binding using passive adsorption techniques.
  • Polybead® Microspheres are monodisperse polystyrene microspheres available in sizes from 50 nm to 90 μm
  • Diameters 4.5 μm and larger are crosslinked with divinylbenzene (DVB)
  • These particles contain a slight anionic charge from sulfate ester
  • Functionalized versions are available for covalent binding
Experienced chemists control the synthesis to provide precise monodisperse particle size distributions. Most Polybead® Microspheres are packaged in anMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable