Stain Kits and Replacement Components, Epredia
Supplier: Epredia
87016 88006 88009 87008 88024 87009 88035 87101 88003 88117 87017 88008 88012 88101 88104 87006 88015 88018 87019 88028 88038 88037 87015 88001 88007 88102 88103 88013 88014 88023 88025 88033 88034 88019 88020 88022 88029 87020 87021 88031 88002 87003 88004 88005 88000 88011 87018 87007 88017 88026 88027 88036 88039 87012 88030
84000-208EA 387.6 USD
84000-208 84000-218 84000-226 84000-260 84000-264 84000-274 84000-284 16002-128 84000-204 84000-206 84000-220 84000-224 84000-234 84000-236 84000-242 84000-244 84000-250 84000-258 84000-286 84000-294 84000-298 84000-306 84000-200 84000-202 84000-222 84000-238 84000-240 84000-246 84000-248 84000-262 84000-266 84000-280 84000-282 84000-288 84000-290 84000-292 84000-296 84000-308 84000-312 84000-314 84000-210 84000-212 84000-214 84000-216 84000-228 84000-230 84000-232 84000-252 84000-256 84000-268 84000-270 84000-272 84000-300 84000-304 84000-310
Stain Kits and Replacement Components, Epredia
Stains Histology Stains
Epredia™ stain kits and replacement components contain all the necessities of the staining procedure.
  • Save time
  • Components available separately
  • All the necessary stains, solutions and components
Acid Fast Bacillus, blue secondary stain kits stain bright red with a pale blue background. Acid Fast Bacillus, green secondary stain kit stains dark red with a pale green background. Demonstrate amyloid with Epredia™ Amyloid Stain. Amyloid stain kit stains orange to red.
Demonstrate the atrophy, thinning and loss of elastic tissue with Epredia™ Elastic Stain. Elastic stain kits stain blue-black to black for elastic tissue, red for collagen with a yellow background.
Save time with Epredia™ Gomori Trichrome (Blue Collagen), a one-step trichrome stain that achieves color differentiation similar to the Masson Trichrome. Gomori Trichrome (Blue Collagen) stain kit stains red for Muscle tissue and cytoplasm, blue for collagen and blue-black for nuclei. Epredia™ Gomori Trichrome (Green Collagen is a one-step trichrome stain that provides distinctive red muscle fibers with green collagen staining.
Detect the presence of ferric iron (hemosiderin) in histological or hematological preparations with Epredia™ Iron Stain. Iron stain kits stain blue iron, pale pink cytoplasm and pink-red nuclei.
Distinguish fungi, basement membranes and opportunistic organisms such as Pneumocystis Carinii in tissue sections with Epredia™ Modified Grocott's Methenamine Silver (Periodic Acid), a uniquely modified silver stain formulation that utilizes methenamine/borax capsules to make working silver solution. Modified Grocott’s Methenamine kit stains black with a green background.
Identify primary tumor sites with the aid of Epredia™ Mucicarmine. Mucicarmine stain kit stains dark rose for mucin and black for nuclei with a yellow background.
Demonstrate the patterns and presence of specific carbohydrates with Epredia™ Periodic Acid-Schiff (PAS) Solution. Periodic Acid Schiff kit stains rose to magenta with blue nuclei.
Determine the presence of spirochetes and nonfilamentous bacteria with Epredia™ Steiner-Steiner Silver. Steiner-Steiner silver kits stain black with a yellow to brown background.
Información para pedidos: All kits come with all stains and solutions necessary to complete the staining procedure. Components of kits are also available separately.
Order Now
SPECIFICATIONS
Environmentally Preferable