VWR® Centrifugal Filters
Supplier: VWR International
516-0229 516-0230 516-0232 516-0227 516-0233 516-0235 516-0236 516-0228 516-0234
82031-348PK 450.23 USD
82031-348 82031-350 82031-354 82031-344 82031-356 82031-360 82031-362 82031-346 82031-358
VWR® Centrifugal Filters
Filters Centrifugal Filters
Use to: Concentrate, purify, and desalt peptides, proteins, oligonucleotides, DNA, and RNA; clean up labeling and PCR reactions; isolate DNA from agarose gel slices; buffer exchange; separate proteins, oligonucleotides, and RNA from acrylamide gels; clarify samples; or remove particulates.
  • Polypropylene housing
  • Membrane options available
Filter housings are constructed of polypropylene and are available with a polyethersulfone (PES) or nylon membrane.
Filters with nylon membrane are ideal for prefiltration clean-up of large agarose or acrylamideMore Product Information