Operating Scissors, Sklar
Supplier: Sklar
13-1045 13-1050 13-1055 13-1060 13-1065 13-1072 13-2045 13-2050 13-2055 13-2060 13-2065 13-2072 14-1045 14-1050 14-1055 14-1060 14-1065 14-1072 14-2045 14-2050 14-2055 14-2060 14-2065 15-1045 15-1050 15-1055 15-1060 15-1065 15-2045 15-2050 15-2055 15-2060 15-2065 22-1145 22-1150 22-1155 22-1245 22-1250 22-1255 22-1345 22-1350 22-1355 22-1445 22-1450 22-1455 22-1550 22-1650 22-1655 22-1545 22-1555 22-1645
82029-884EA 77.91 USD
82029-884 82029-886 82029-888 82029-890 82029-892 82029-894 82029-896 82029-898 82029-900 82029-902 82029-904 82029-906 82029-908 82029-910 82029-912 82029-914 82029-916 82029-918 82029-920 82029-922 82029-924 82029-926 82029-928 82029-932 82029-934 82029-936 82029-938 82029-940 82029-948 82029-950 82029-952 82029-954 82029-956 89025-214 89025-216 89025-218 89025-236 89025-238 89025-240 89025-244 89025-246 89025-250 89025-254 89025-256 89025-258 89025-264 89025-274 89025-276 10195-204 10195-206 10195-208
Operating Scissors, Sklar
Scissors
Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument.
With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures, and multiple lengths, these scissors are used for general, multi-purpose cutting, and dissecting
Censitrac Ready OR Grade Operating Scissors are industry standard and constructed of stainless steel.
Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze.