FOCUS™ Soluble and Insoluble, G-Biosciences
Supplier: G-Biosciences
786-247
82022-672EA 672.64 USD
82022-672
FOCUS™ Soluble and Insoluble, G-Biosciences
Protein Extraction Protein Extraction Kits
A complete kit for the selective preparation of soluble (hydrophilic) and insoluble (hydrophobic) proteins from mammalian tissues and cells, plants, yeast, bacteria, and other biological samples
Also supplied with a specific clean-up kit for the preparation of each fraction for isoelectric focusing and 2D electrophoresis for improved spot resolution. FOCUS™ Soluble & Insoluble kit is designed for 50 preps, where 1 prep is one of the following: 100mg mammalian tissue, 50μl wet animal cell pellet, 50μL wet yeast pellet, 50μL wet bacteria pellet, or 250 mg plant tissue.
Supplied with reagents necessary for fractionation of soluble and insoluble fractions, including a strong chaotropic extraction buffer to solubilize difficult proteins.
Información para pedidos: Accessories available.