VWR® Electronic Stem Thermometer, Calibrated, Stainless Steel, 197 mm Probe
Supplier: VWR International
82021-164
82021-164EA 106.59 USD
82021-164
VWR® Electronic Stem Thermometer, Calibrated, Stainless Steel, 197 mm Probe
Thermometers Digital Thermometers
Digital stem thermometer is suitable for field studies, wet lab applications, wash down areas, food testing and tanks.
  • Blunt tip probe is ideal for veterinary practices, farm and ranch
  • Will read temperatures in liquids, semi-solids and gases
  • Stainless-steel probe easily reaches into cuvettes, test tubes, flasks, beakers and more
  • Individually serialized, CE marked
Certificaciones: Tested and calibrated in exclusive triple accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration isMore Product Information