Removable Large Hub Needles for Syringes 250 μl to 10 ml, Hamilton Company
Supplier: Hamilton
7780-03 7780-04 7779-02 7779-01 7779-03 7779-04 7780-01 7780-02 7784-04 7784-03 7784-01 7784-02 7732-02
82007-206PK 87.36 USD
82007-206 82007-248 82007-736 82010-264 82010-268 82010-270 82010-272 82010-274 82010-276 82010-278 82010-282 83007-502 89220-924
Removable Large Hub Needles for Syringes 250 μl to 10 ml, Hamilton Company
Syringe Needles
Hamilton Company offers the most comprehensive selection of standard and custom needles in the industry. Each is manufactured to achieve the highest level of accuracy and precision possible.
Removable Needle (RN)- The needles are removable and are a Hamilton-specific design
Needle Point Styles (Custom gauges, lengths and point styles available):
Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 34-10.
Point Style 3 - Blunt needle point for use with HPLC injection valves and for sample pipetting. Available gauges: 34-10.
More Product Information