Thermoplastic Duct Reducers, Labconco®
Supplier: Labconco
5605900 5606000 5606100 5630700
82006-934EA 330 USD
82006-934 82006-936 82006-938 82006-940
Thermoplastic Duct Reducers, Labconco®
Ducting Supplies
PVC coupling type reducers are designed for connecting thermoplastic duct of different diameters
Compare your blower inlet size with your duct size to see if one is necessary.
Order Now
SPECIFICATIONS
Environmentally Preferable