Heidolph® Reax Top and Reax Control Vibrating Shakers
Supplier: Heidolph NA, LLC
036130000 036130020
82004-930EA 814.79 USD
82004-930 82004-932
Heidolph® Reax Top and Reax Control Vibrating Shakers
Shakers and Mixers Orbital Shakers
For vigorous shaking of small vessels.
The small test tube tray holds up to 20 ml test tubes and flasks. The large test tube tray holds up to 50 ml test tubes and flasks. The foam attachment provides space for the simultaneous mixing of up to 10 test tubes.
The Reax Top model is a standard test tube shaker with analog dial speed control. The Reax Control model provides constant vibration under changing loads with electric speed control. Shaker units feature a sparkless, maintenance-free motor and sealed housing to prevent moisture and splashes from interfering with performance. UnitsMore Product Information