Vibration Isolation Table, Labconco®
Supplier: Labconco
3618000
82000-984EA 1685 USD
82000-984
Vibration Isolation Table, Labconco®
Furniture Tables and Workbenches
The vibration isolation table provides an isolated work surface for conducting procedures with vibration-sensitive equipment such as microscopes and balances.
  • Four leveling feet
  • Work surface is laminated hard board
The table never makes contact with the clean bench, so vibration from the motor/blower is not transmitted to the table. Epoxy-coated steel frame, adjusts in 1" increments to provide a working height from 29.5 to 36". ADA-compliant. Compatible with Purifier Horizontal Clean Benches.