DNA sodium salt (Deoxyribonucleic acid sodium salt) (from salmon testes)

Supplier: MilliporeSigma

Synonyms: Thymonucleic acid sodium salt, DNA sodium salt

262012-1GM 262012-5GM
80601-118EA 146.05 USD
80601-118 80601-120
DNA sodium salt (Deoxyribonucleic acid sodium salt) (from salmon testes)
DNA sodium salt (Deoxyribonucleic acid sodium salt)

Purified DNA from salmon testes.


  • N/P ratio: 1.5 to 1.8
  • Absorption (0.001% solution in phosphate buffer, pH 7.0, 260 nm): ≥0.155
  • Storage: 2 to 8 °C
  • Form: White to off-white to light brown solid
  • Hygroscopic

MDL Number: MFCD00130921
CAS Number: 9007-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR