B.O.D. Bottles, Wheaton, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
227494-00 227494-99 227494-03 227494-00G 227494-99G 227494-01 227494-02
16286-070CS 2162.55 USD
16286-070 74830-044 74830-048 74830-050 80093-740 80093-742 80093-744 80093-746 80093-748 80093-750
B.O.D. Bottles, Wheaton, WHEATON®, DWK Life Sciences
Bottles BOD Bottles
Ideal for incubating diluted samples of sewage, sewage effluents, polluted waters, and industrial wastes.
These 60 ml borosilicate glass bottles feature a shoulder radius that sweeps all air from inside the bottle during filling.
The bottles are designed with a permanently screened-on label and are available with either glass robotic stoppers or glass pennyhead stoppers. Bottles may be used with B.O.D. bottle rack. Bottle rack is rugged with plastic coating for corrosion resistance and quietness and holds twenty 60 ml B.O.D. bottles.
The interchangeable stoppers have a taperedMore Product Information