GreenHouse Blowdown Evaporator, System 16, Heidolph Radleys
Supplier: Heidolph NA, LLC
015850160
80086-234EA 11678.7 USD
80086-234
GreenHouse Blowdown Evaporator, System 16, Heidolph Radleys
Evaporators Parallel Synthesis Systems
Turn your existing GreenHouse Parallel Synthesizer into a parallel evaporator, or simply use this system as your own personal benchtop evaporator
The GreenHouse Blowdown uses a precisely controlled flow of inert gas combined with digitally controlled heating to carefully evaporate your samples. Interchangeable plates with either 8 or 24 hollow pins deliver an equal flow of gas to each tube, vial or well. The absence of any vacuum avoids bumping, further protection you sample during evaporation.
Información para pedidos: This System includes GreenHouse Blowdwon head, Long Blowdown pinMore Product Information