Potassium chloride 3 M in aqueous solution

Supplier: YSI INC

Synonyms: Potassiumchloride

109706
80085-324EA 41.78 USD
80085-324
Potassium chloride 3 M in aqueous solution
Potassium chloride
Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1420
Melting Pt: 773
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR