Glas-Col® Mantle Extension Supports, Series M, Chemglass
Supplier: Chemglass
CG-11004-02 CG-11004-03 CG-11004-04 CG-11004-05
80060-360EA 216.6 USD
80060-360 80060-362 80060-364 80060-366
Glas-Col® Mantle Extension Supports, Series M, Chemglass
Heaters Heating Mantles
Glas-Col® extension support for use with a smaller Series M mantle.
  • Attached to a stable ring stand or distillation rack
  • The support can be adjusted to any convenient height
  • Support is painted gray and has a 6" long, ½" O.D. rod