Dodecyl-β-D-glucopyranoside

Supplier: MilliporeSigma

Synonyms: Dodecyl b-D-glucopyranoside, Dodecyl glucoside, n-Dodecyl β-D-glucopyranoside

324351-1GM
80056-362EA 310.53 USD
80056-362
Dodecyl-β-D-glucopyranoside
Dodecyl-β-D-glucopyranoside
Formula: C₁₈H₃₆O₆
MW: 348.48 g/mol
MDL Number: MFCD00063298
CAS Number: 59122-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR