NAD+ free acid ≥95% (by HPLC), Calbiochem®, Millipore®

Supplier: MilliporeSigma

Synonyms: 1-((2R,3R,4S,5R)-5-((((((((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)oxidophosphoryl)oxy)methyl)-3,4-dihydroxytetrahydrofuran-2-yl)-3-carbamoylpyridin-1-ium

481911-5GM
80053-322EA 440.73 USD
80053-322
NAD+ free acid ≥95% (by HPLC), Calbiochem®, Millipore®
NAD+

NAD⁺, Free Acid, a major electron acceptor molecule in biological oxidations. Spectra (pH 7.5): 250/260 nm: 0.80-0.86; 280/260 nm: 0.19-0.23; 340/260 nm: 0.42-0.44 (with ADH at pH 10).

Formula: C₂₁H₂₇N₇O₁₄P₂
MW: 663.425 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00036253
CAS Number: 53-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR