Daidzein, Calbiochem®

Supplier: MilliporeSigma

Synonyms: Daidzeol, 7-Hydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 7-Hydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',7-Dihydroxyisoflavone, 7-Hydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one

251600-25MG
80051-320EA 85.81 USD
80051-320
Daidzein, Calbiochem®
Daidzein

Inactive analog of genistein


Blocks the G1 phase of the cell cycle in Swiss 3T3 cells by inhibiting casein kinase II activity.

Formula: C₁₅H₁₀O₄
MW: 254.24 g/mol
Boiling Pt: 513
MDL Number: MFCD00016954
CAS Number: 486-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR