BCECF AM ester ≥85% (by HPLC), Calbiochem®, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 2′,7′-Bis(2-carboxyethyl)-5(6′)-carboxyfluorescein acetoxymethyl ester, Spiro(isobenzofuran-1(3H),9'-(9H)xanthene)-2',7'-dipropanoic acid, BCECF AM

M-Clarity™ quality level = MQ100

216254-1MG
80050-710EA 247.97 USD
80050-710
BCECF AM ester ≥85% (by HPLC), Calbiochem®, Sigma-Aldrich®
BCECF AM ester

A variable mixture of cell-permeable ester derivatives of BCECF that are hydrolyzed by cytosolic esterases to yield intracellularly trapped indicator BCECF. Has been used to monitor intracellular pH changes in mammalian fibroblasts, gastric cells, lymphocytes, myocytes, and distal convoluted tubule


Envase: Packaged under inert gas

Formula: C₄₂H₄₀O₂₁
MW: 808.69 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00036969
CAS Number: 117464-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR