HMS174 Competent Cells, Expression Strains, MilliporeSigma
Supplier: MilliporeSigma
69453-4 70234-3 69454-3 69452-3 69452-4 69454-4 69453-3
80030-336EA 453.51 USD
80030-336 80030-832 80030-338 80030-330 80030-332 80030-340 80030-334
HMS174 Competent Cells, Expression Strains, MilliporeSigma
Competent Cells
A common method for transformation of plasmids into E.
DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lac UV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability.
The standard volume per tube is 0.2 ml,More Product Information