Vikan® Table and Floor Scrapers
Supplier: Remco Products
29082 29083 29084 29085 29086 29087 29089
77906-512EA 28.9 USD
77906-512 77906-494 77906-504 77906-518 77906-496 77906-506 77906-500
Vikan® Table and Floor Scrapers
Scrapers
Remove/loosen stubborn food debris or labels from floors and other surfaces with this table and floor scraper.
  • Durable and solid blade
  • Threaded connection
The table and floor scraper features a very durable and solid stainless-steel blade within a polypropylene housing. This scraper can be used with any threaded Vikan® handle. An ideal fit for any color-coded cleaning systems.
Certificaciones:
FDA compliant.