Refrigerated Incubators
Supplier: Thermo Fisher Scientific
TFSRI150-115V TFSRI250-115V TFSRI420-115V TFSRI720-115V
77889-372EA 6000 USD
77889-372 77889-376 77889-380 77889-384
Refrigerated Incubators
Incubators Cooled Incubators
Thermo Scientific Refrigerated Incubators utilize robust compressor cooling to help ensure a controlled temperature environment to fit your application needs.
  • 115 V models
  • Temperature range 4 to 60 °C
  • Temperature stability and uniformity fitting most applications
  • Environmentally-friendly hydrocarbon refringent, R290, has a Global Warming Potential index (GWP) of three, keeping the environmental impact of the refrigeration system minimized
  • 7" touch screen for data export, alarm history, operating status, and real-time temperature curve
  • Coated cabinet
More Product Information