2-Thiopheneboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 4,4,5,5-Tetramethyl-2-(2-thienyl)-1,3,2-dioxaborolane

A261566-100G A261566-10G A261566-25G A261566-5G A261566-1G A261566-500G
103812-528EA 10 USD
103812-528 103852-418 76692-716 76692-718 77092-022 77092-024 77304-816 77879-489
2-Thiopheneboronic acid pinacol ester 97%
2-Thiopheneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₀H₁₅BO₂S
MW: 210.1 g/mol
Boiling Pt: 286.5 °C (760 mm Hg)
Melting Pt: 68…70 °C
Flash Pt: 127.099 °C
Storage Temperature: Freezer
MDL Number: MFCD05663878
CAS Number: 193978-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR