Methyl chrysin,Tectochrysin ≥98%

Supplier: Ambeed
Warning

Synonyms: Tectochrysin, 5-Hydroxy-7-methoxy-2-phenyl-4H-chromen-4-one

A956987-100MG
77878-756EA 237 USD
77878-756
Methyl chrysin,Tectochrysin ≥98%
Methyl chrysin,Tectochrysin

Precaución: for Research Use Only

Formula: C₁₆H₁₂O₄
MW: 268.27 g/mol
MDL Number: MFCD00016945
CAS Number: 520-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR