Benzo-15-crown-5 (benzo-1,4,7,10,13-pentaoxacyclopentadecane) 98%

Supplier: Ambeed
Warning

Synonyms: Benzo-15-crown-5

A120203-1G A120203-5G A120203-100G A120203-25G
76688-074EA 10 USD
76688-074 76688-076 77566-220 77876-873
Benzo-15-crown-5 (benzo-1,4,7,10,13-pentaoxacyclopentadecane) 98%
Benzo-15-crown-5 (benzo-1,4,7,10,13-pentaoxacyclopentadecane)

Precaución: For research use only.

Formula: C₁₄H₂₀O₅
MW: 268.31 g/mol
MDL Number: MFCD00005945
CAS Number: 14098-44-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR