Orion™ AQUAfast COD165 Thermoreactor
Supplier: Thermo Fisher Scientific
COD165
77870-936EA 1650 USD
77870-936
Orion™ AQUAfast COD165 Thermoreactor
Block Heaters Digital Block Heaters
Conduct environmental and wastewater monitoring with the Thermo Scientific™ Orion™ AQUAfast COD165 Thermoreactor. The COD165 thermoreactor is equipped with an automatic voltage selector for voltages between 90 and 260 V with a frequency of 50 or 60 Hz. Select from five preset work temperatures of 100, 120, 150, 160, 165 °C and four preset run times of 30, 60, 120 minutes or continuous time. Simply place 16 mm round sample tubes into the digestor wells, cover with the hinged safety shield, set the temperature and time and let samples digest.
  • Temperature settings for 100, 120, 150, 160
More Product Information