Brady® i75 R6300 Ribbon for the i7500 Label Printer
Supplier: Brady Worldwide
176832 176833 176834 176835
77775-104RL 0 USD
77775-104 77775-106 77775-108 77775-110
Brady® i75 R6300 Ribbon for the i7500 Label Printer
Printer Ribbons
i75-Series ribbon with LabelSense™ Technology warns user if ribbon is incompatible with installed i75-Series label material. Automatically prints at pre-tested heat and speed, ensuring correct print quality on the first label with no setting adjustments or trial-and-error print testing.
  • Ribbon in cartridge format
  • Ribbin ink wind direction: ink side out
  • Polyester material
  • Halogen-free
i75-series ribbon is self-centering with ink-side always correctly facing the label. One-color ribbon for printing a single color onto a label. R6300 Series resin
More Product Information