Brady® i75 Series R6000 Ribbon for the i7500 Label Printer
Supplier: Brady Worldwide
176752 176753 176754 176755
77775-028RL 0 USD
77775-028 77775-030 77775-032 77775-034
Brady® i75 Series R6000 Ribbon for the i7500 Label Printer
Printer Ribbons
i75-series ribbon with LabelSense™ Technology warns user if ribbon is incompatible with installed i75-series label material. Automatically prints at pre-tested heat and speed, ensuring correct print quality on the first label with no setting adjustments or trial-and-error print testing.
  • Ribbon in cartridge format
  • Ribbon ink wind direction: Ink side out
  • Polyester material
  • Halogen-free
i75-series ribbon is self-centering with ink-side always correctly facing the label. One-color ribbon for printing a single color onto a label. R6000 Series halogen-free
More Product Information