Brady® i75 Series R4900 Ribbon for the i7500 Label Printer
Supplier: Brady Worldwide
176750 176751
77775-024RL 0 USD
77775-024 77775-026
Brady® i75 Series R4900 Ribbon for the i7500 Label Printer
Printer Ribbons
i75-series ribbon with LabelSense™ Technology warns user if ribbon is incompatible with installed i75-series label material. Automatically prints at pre-tested heat and speed, ensuring correct print quality on the first label with no setting adjustments or trial-and-error print testing.
  • Ribbon in cartridge format
  • Ribbon ink wind direction: Ink side out
  • Polyester material
  • Halogen-free
i75-series ribbon is self-centering with ink-side always correctly facing the label. One-color ribbon for printing a single color onto a label. R4900 Series resinMore Product Information