Anti-RSV Human Recombinant Monoclonal Antibody [clone: RV11]

Supplier: ReVacc Scientific
L46X29-100UG L46X29-20UG
77761-684EA 997.02 USD
77761-684 77761-686
Anti-RSV Human Recombinant Monoclonal Antibody [clone: RV11]
Antibodies
This antibody has been tested to bind to fusion (F) glycoprotein of human Respiratory Syncytial Virus (RSV) subtype A1 strain (Cat. No. 77761-674) or B1 strain (Cat. No. 77761-676). An example of comparison of IgM and IgG using size exclusion chromatography is shown.

Type: Primary

Antigen: RSV

Clonality: Recombinant Monoclonal

Clone: RV11

Conjugation: Unconjugated

Epitope:

Host: Human

Isotype: IgM kappa

Reactivity: Human


New Product
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR